PCR plates, 96-well, segmented, semi-skirted, barcoded
Supplier: Thermo Fisher Scientific
Certificates
About this item
Proprietary segmented plate design allows plates to be cut into 24- and 48-well sections. These plates are made from PP.
- White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
- Cut corner: H1
- Raised rim design around each well enables secure sealing and safeguards against evaporation
- Uniform, thin well walls deliver maximum and consistent heat transfer
- Semi-skirt adds rigidity and allows for labeling or barcoding
The barcodes are the industry-standard code 128. The label itself can withstand temperatures from -196 up to 120 °C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any "rubbing off" of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.
Frequently Bought Together