Calcium sulphate, Drierite™ 6 mm (4 mesh) for drying
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
CAS Number:7778-18-9
:
Formula:CaO₄S
MDL Number:MFCD00010912
Density:2.964 g/cm³ (20 °C)
Molecular Weight:136.14 g/mol
Melting Pt:1350
Merck Index:14,1706
EINECS:231-900-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together