Order Entry
Czech Republic
ContactUsLinkComponent
Thiocyclam hydrogen oxalate
Thiocyclam hydrogen oxalate
Catalog # EHERC17480000
Supplier:  EHRENSTORFER
CAS Number:  
Thiocyclam hydrogen oxalate
Catalog # EHERC17480000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

 :
EINECS:250-859-2
 :GHS07
CAS Number:31895-22-4
Formula:C₇H₁₃NO₄S₃