Triethylaluminium 25% in toluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Triethylaluminum
CAS Number:97-93-8
:
Formula:C₆H₁₅Al
MDL Number:MFCD00009015
Boiling Pt:128
Molecular Weight:114.17 g/mol
Melting Pt:-46.1
ADR:4.2 I
:M-Clarity™ quality level = MQ100
EINECS:202-619-3
UN:3394
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together