2,5-Dihydrofuran, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1708-29-8
:
Formula:C₄H₆O
MDL Number:MFCD00003219
Molecular Weight:70.09 g/mol
:M-Clarity™ quality level = MQ100
EINECS:216-957-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together