Order Entry
Czech Republic
ContactUsLinkComponent
 
Di-isopropanolamine 99%
 
undefined
Di-isopropanolamine 99%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to light yellow Low melting mass
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4615 to 1.4635 (20°C, 589 nm)
  • Titration with HClO4:
    ≥98.5 %

 

 

Frequently Bought Together