ContactUsLinkComponent
n-Heptane, extra dry 99+%, AcroSeal®
undefined
n-Heptane, extra dry 99+%, AcroSeal®
  • Environmentally Preferable:
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.0005 %
Frequently Bought Together