Ammonium acetate, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Acetic acid ammonium salt
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
Density:1.17 g/cm³ (20 °C)
Molecular Weight:77.08 g/mol
Melting Pt:114
Flash Pt:136
Merck Index:13,00495
:M-Clarity™ quality level = MQ200
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:1480 g/l (4 °C)
- Melting Point:114 °C
- Molar Mass:77.08 g/mol
- Bulk Density:410 kg/m3
- Density:1.17 g/cm3 (20 °C)
- pH:6.7 - 7.3 (50 g/l, H2O, 25 °C)
- Storage:Store at +15°C to +25°C.
- Hill Formula:C2H7NO2
- Chemical Formula:CH3COONH4
Specification Test Results
Frequently Bought Together