Butyric acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Butanoic acid
CAS Number:107-92-6
:
Formula:C₄H₈O₂
MDL Number:MFCD00002814
Boiling Pt:162…164 °C
Melting Pt:–8 °C
Density:0.964
Molecular Weight:88.11 g/mol
Flash Pt:69 °C (156 °F)
UN:2820
ADR:8 III
Merck Index:13,01593
EINECS:203-532-3
:GHS06
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberALFAL13189.AE_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together