Erlenmeyer flasks, narrow neck, with flared rim
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
White graduations and labelling area.
: ISO 1773
Frequently Bought Together