Order Entry
Czech Republic
ContactUsLinkComponent
Oxiracetam 97%
Oxiracetam 97%
New Product
Certificates
Oxiracetam 97%
New Product

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    white to almost white crystalloine powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Melting point:
    169°C to 173°C
  • Loss on drying:
    ≤3 %

Specification Test Results

Frequently Bought Together