Troclosene sodium 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium dichloroisocyanurate, Dichloro-1,3,5-triazinetrione sodium salt, Dichloroisocyanuric acid sodium salt
CAS Number:2893-78-9
:
Formula:C₃Cl₂N₃NaO₃
MDL Number:MFCD00006036
Molecular Weight:219.95 g/mol
Melting Pt:225
Storage Temperature:Ambient
EINECS:220-767-7
UN:2465
ADR:5.1 II
:GHS03
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystals or granules
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Titration with HClO4:≥97.5 % (On dry substance)
Specification Test Results
Frequently Bought Together