Anti-TIMP2 Mouse Monoclonal Antibody [clone: TIMP2/2044]
ANTIA253315-100
New Product
- Antibody type:Primary
- Antigen name:CSC 21K
- Antigen symbol:TIMP2
- Clonality:Monoclonal
- Clone:TIMP2/2044
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P16035
- Antigen synonyms:DDC8|MGC105282|OTTMUSP00000004171|CSC21K|CSC-21K|Metalloproteinase inhibitor 2 precursor|OTTMUSP00000004170|D11Bwg1104e|Metalloproteinase inhibitor 2
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:21 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 48 - 220, of human TIMP2 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [TIMP2/2044] antibody to TIMP2 for ELISA and IHC-P with samples derived from human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: TIMP2/2044
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human