Anti-C1 inh Mouse Monoclonal Antibody [clone: KT28]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Mouse monoclonal (KT28) antibody to C1 inh for ELISA with samples derived from human.
- Validated applications: ELISA
- Recommended dilutions: ELISA: 1 µg/ml
Type: Primary
Antigen: C1 inh
Clonality: Monoclonal
Clone: KT28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ANTIA47-100
- ANTIA47-200
- Antigen symbol:
- C1 inh
- C1 inh
- Antigen name:
- C1 esterase inhibitor
- C1 esterase inhibitor
- Antigen synonyms:
- C1-inhibiting factor|C1 Inh|C1IN|complement component 1 inhibitor|C1 inhibitor|C1Inh|C1 Inactivator|C1 inhibiting factor|C1NH
- C1-inhibiting factor|C1 Inh|C1IN|complement component 1 inhibitor|C1 inhibitor|C1Inh|C1 Inactivator|C1 inhibiting factor|C1NH
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- KT28
- KT28
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P05155
- UniprotID# P05155
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Immunogen:
- Purified C1 inhibitor from human blood
- Purified C1 inhibitor from human blood
- Form:
- Liquid
- Liquid
- Purification:
- IgG is purified through a Protein A column.
- IgG is purified through a Protein A column.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together