Gabexate mesylate ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxy]benzoic acid ethyl ester Mesylate, Ethyl 4-((6-guanidinohexanoyl)oxy)benzoate methanesulphonate, Gabexate Methanesulphonate
CAS Number:56974-61-9
:
Formula:C₁₆H₂₃N₃O₄·CH₃SO₃H
MDL Number:MFCD00210299
Molecular Weight:417.5 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together