Phosphotungstic acid 44-hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Trioxotungsten - phosphoric acid hydrate (12:1:44), Tungsten, trioxo-, compd. with phosphoric acid, hydrate (12:1:44)
CAS Number:12067-99-1
Formula:H₉₁O₈₄PW₁₂
MDL Number:MFCD00149916
Molecular Weight:3672.73 g/mol
Storage Temperature:Ambient
EINECS:235-087-6
UN:3260
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together