Low retention filter pipette tips, ZEROTIP®
JET ZEROTIP® are manufactured from super-clear high quality polypropylene and are compatible with the majority of different micropipette models and brands.
- Preferred accessories for most brands of micropipettes
- Colour-coded racks for easy volume identification
- Every rack or case is printed with lot no. for quality traceability
- DNase-/RNase-free and non pyrogenic
- Sterilised by irradiation SAL 10⁻⁶ (ISO 11137)
- Shelf life: 3 years after month of production
The surfaces of the tips are produced through a special process. This process makes the tip inner surface become super hydrophobic, thus significantly reduces sample loss, and provides substantially higher reproducibility when working with critical media.
: Manufactured in a Class 100 000 cleanroom environment. Manufactured under EN ISO 13485:2016 and ISO 9001:2015 quality management systems.
- Catalog No:
- 732-3699
- 732-3697
- 732-3693
- 732-3703
- 732-3701
- 732-3705
- 732-3707
- 732-3695
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Frequently Bought Together