2-Chloro-4-fluoro-5-nitrophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84478-75-1
:
Formula:C₆H₃ClFNO₃
MDL Number:MFCD02670258
Molecular Weight:191.5400 g/mol
Melting Pt:105…108 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together