1-Methylcyclopropanamine hydrochloride 98%
Catalog # APOSOR16691-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:107.5800 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:88887-87-0
Formula:C₄H₁₀ClN
MDL Number:MFCD11109576
Melting Pt:196…205 °C