4-Chlorothiazol-2-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52107-46-7
:
Formula:C₃H₃ClN₂S
MDL Number:MFCD09999189
Molecular Weight:134.587 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together