2-Nitro-5-thiocyanatobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Nitro-5-Thiocyanobenzoic acid
CAS Number:30211-77-9
:
Formula:C₈H₄N₂O₄S
MDL Number:MFCD00007139
Molecular Weight:224.2 g/mol
Melting Pt:162…163 °C
:M-Clarity™ quality level = MQ200
EINECS:250-093-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together