Sodium sulphide hydrate 60 - 64%, scales, extra pure
: di-Sodium sulphide hydrate
:27610-45-3
:
:Na₂S·xH₂O
:MFCD00149183
:920
:96.06 g/mol (anhy)
:50
:8 II
:12,08829
:215-211-5
:1849
:01-2119513694-38
:GHS05
:GHS06
:GHS09
:Refrigerator
- Environmentally Preferable:
- Appearance:Yellow Flakes
- Titration Iodimetric:60 to 64 % (Na2S)
- Sodium carbonate (Na2CO3):≤1.2 % (Na2CO3)
- Iron (Fe):≤10 ppm
Frequently Bought Together