Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyrrole
Pyrrole ≥98%
Pyrrole ≥98%
:
Thermo Fisher Scientific
Pyrrole ≥98%
:
Thermo Fisher Scientific
:
1H-Pyrrole
:
109-97-7
:
:
C₄H₅N
:
MFCD00005216
:
131 °C
:
–24 °C
:
0.967
:
67.09 g/mol
:
38 °C (100 °F)
:
1992
:
3 III
:
12,08198
:
203-724-7
:
GHS06
:
GHS05
:
GHS02
Environmentally Preferable:
msds_ALFAA12616
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
N,N-Dimethylformamide, EMSURE® ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Hexane 95, PESTINORM® Pest_Residual
: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99,5, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Reag. USP, ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol 70% 70 v/v, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol 99,0, GPR RECTAPUR®
: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® LC-MS
: VWR Chemicals