Mercury(II) sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Mercury sulphate, Mercuric sulphate
CAS Number:7783-35-9
:
Formula:HgSO₄
MDL Number:MFCD00011047
Density:6.47 g/cm³ (25 °C)
Molecular Weight:296.65 g/mol
Melting Pt:850
ADR:6.1 II
Merck Index:13,05914
EINECS:231-992-5
UN:1645
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together