Pyrrolidine 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tetrahydropyrrole
CAS Number:123-75-1
:
Formula:C₄H₉N
MDL Number:MFCD00005249
Boiling Pt:86…88 °C (760 mmHg)
Melting Pt:–63 °C
Density:0.866 g/cm³
Molecular Weight:71.12 g/mol
Flash Pt:3 °C
Storage Temperature:Ambient
UN:1922
ADR:3 II
Merck Index:12,08199
EINECS:204-648-7
:GHS07
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4421 to 1.4441 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥99.0 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together