Acetic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118
Melting Pt:17
Density:1.05 g/cm³ (20 °C)
Molecular Weight:60.05 g/mol
Flash Pt:38.5
Storage Temperature:Ambient
UN:2789
ADR:8 II
:M-Clarity™ quality level = MQ300
EINECS:200-580-7
REACH:01-2119475328-30
:GHS02
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together