L-Arginine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
CAS Number:1119-34-2
:
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550
Density:1.42 g/cm³ (20 °C)
Molecular Weight:210.66 g/mol
Melting Pt:218
Merck Index:13,00785
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:214-275-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together