Xylometazoline hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(4-(tert-Butyl)-2,6-dimethylbenzyl)-4,5-dihydro-1H-imidazole hydrochloride, 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline Hydrochloride
CAS Number:1218-35-5
:
Formula:C₁₆H₂₄N₂
MDL Number:MFCD00238707
Molecular Weight:280.84 g/mol
UN:2811
ADR:6.1 III
:GHS06
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 4 h)
- Solubility:(5 % in water) Clear colorless to light yellow
- Titration with HClO4:99 to 101 % (dry substance)
Specification Test Results
Frequently Bought Together