Vacuum greases, Apiezon®
These Apiezon greases are hydrocarbon based, silicone-free and highly resistant to creep, the undesirable phenomenon associated with silicone-based products.
- Excellent performance in a wide range of vacuum applications
- Silicone-free, highly creep resistant
- Solutions for a variety of temperatures
- Stable, chemically inert and non toxic
- Easy to use and remove
Using Apiezon reduces the chance of grease travelling away from the area of application and contaminating adjacent surfaces. In addition the ultra high vacuum grease ensures ultimate cleanliness and low outgassing without contaminating vacuum systems.
Apiezon L Grease and Apiezon M Grease are halogen-free products and therefore has a built-in resistance to irradiation.
- Catalog No:
- 24491.133
- 24488.154
- 24487.184
- MIMAAP100-0050
- 24487.138
- 24486.157
- 24486.135
- MIMAFAPGAP10104000
- Temperature range:
- −10...+240 °C
- −40...+180 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- −40...+180 °C
- Type:
- Apiezon® H
- Apiezon® AP101
- Apiezon® M
- Apiezon® AP100
- Apiezon® M
- Apiezon® L
- Apiezon® L
- Apiezon® AP101
- Vapor pressure:
- 1,7 x 10-9 Torr (20 °C)
- <1.0 x 10-5 Torr (20 °C)
- 7 x 10-9 Torr (20 °C)
- 7 x 10-11 Torr (20 °C)
- 7 x 10-9 Torr (20 °C)
- 7 x 10-11 Torr (20 °C)
- 7 x 10-11 Torr (20 °C)
- <1.0 x 10-5 Torr (20 °C)
Frequently Bought Together