About this item
This benchtop meter is ideal for a wide range of applications and advanced pH and conductivity analysis in the lab. It offers two measuring channels allowing users to measure pH and conductivity simultaneously or view each channel separately.
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Non volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.
Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two.
Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values. Perform up to a five point conductivity calibration. Calibration editing option allows individual points to be fixed without a full recalibration. Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate.
Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.
The kit additionally includes: 8157BNUMD ROSS Ultra Triode refillable epoxy-body pH/ATC electrode, 013005MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor, ROSS pH buffer and storage solution kit and Orion 1413 µS conductivity standard (5× 60 ml).
Specifications
- Catalog No:
- 665-0272
- 665-0271
- Model:
- A215
- A215
- pH range:
- –2,000...+20,000
- –2,000...+20,000
- pH resolution:
- 0,1 / 0,01 / 0,001
- 0,1 / 0,01 / 0,001
- pH accuracy:
- ±0,002
- ±0,002
- pH calibration:
- Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- mV range:
- ±2000,0
- ±2000,0
- mV resolution:
- 0,1
- 0,1
- mV accuracy:
- ±0,2 mV or ±0,05% of reading, whichever is greater
- ±0,2 mV or ±0,05% of reading, whichever is greater
- Conductivity range:
- 0,001 µS/cm - 3000 mS/cm
- 0,001 µS/cm - 3000 mS/cm
- Conductivity resolution:
- 0,001 μS minimum, auto ranging, up to 4 significant digits
- 0,001 μS minimum, auto ranging, up to 4 significant digits
- Conductivity accuracy:
- 0,5% of reading ±1 digit > 3 μS, 0,5% of reading ±0,01 μS ≤ 3 μS
- 0,5% of reading ±1 digit > 3 μS, 0,5% of reading ±0,01 μS ≤ 3 μS
- Conductivity cell constant:
- 0,001 - 199,9 cm⁻¹
- 0,001 - 199,9 cm⁻¹
- Conductivity calibration:
- Up to 5 points
- Up to 5 points
- Resistivity range:
- 2 Ω/cm - 100 MΩ/cm
- 2 Ω/cm - 100 MΩ/cm
- Resistivity accuracy:
- 0,5% of reading ±1 digit
- 0,5% of reading ±1 digit
- Resistivity resolution:
- 1 ohm or 0,1 meg-ohm, auto ranging
- 1 ohm or 0,1 meg-ohm, auto ranging
- Salinity range:
- 0,06 - 80,00 psu / 0,05 - 42,00 ppt
- 0,06 - 80,00 psu / 0,05 - 42,00 ppt
- Salinity resolution:
- 0,01 psu or 0,01 ppt minimum, auto ranging
- 0,01 psu or 0,01 ppt minimum, auto ranging
- Salinity accuracy:
- 0,5% of reading ±1 digit
- 0,5% of reading ±1 digit
- TDS range:
- 0,001 - 200,0 ppm
- 0,001 - 200,0 ppm
- TDS resolution:
- 0,001 ppm minimum, auto ranging, up to 4 significant digits
- 0,001 ppm minimum, auto ranging, up to 4 significant digits
- TDS accuracy:
- 0,5% of reading ±1 digit
- 0,5% of reading ±1 digit
- Temperature range:
- –5,0...+105,0 °C
- –5,0...+105,0 °C
- Temperature resolution:
- 0,1 °C
- 0,1 °C
- Temperature accuracy:
- ±0,1 °C
- ±0,1 °C
- Temperature compensation:
- Linear, non linear nLFn, non linear nLFu, EP (USP), off
- Linear, non linear nLFn, non linear nLFu, EP (USP), off
- Sensor type:
- DuraProbe 4-cell conductivity sensor
- DuraProbe 4-cell conductivity sensor
- Display:
- Graphic LCD with backlight
- Graphic LCD with backlight
- Battery life:
- 800 h
- 800 h
- Power supply:
- AC adapter or 4 AA batteries
- AC adapter or 4 AA batteries
- Compliance:
- IP 54
- IP 54
- Operating temperature range:
- 5...45 °C (ambient)
- 5...45 °C (ambient)
- Data logging - log function:
- Manual / Automatic with AUTO-READ™
- Manual / Automatic with AUTO-READ™
- Data logging capacity:
- 2000 with date and time stamp
- 2000 with date and time stamp
- IP rating:
- IP 54
- IP 54
- Connections:
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- Outputs:
- RS232, USB
- RS232, USB
- Input:
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- Reference temperature:
- 5 / 10 / 15 / 20 / 25 °C
- 5 / 10 / 15 / 20 / 25 °C
- Alarm:
- High/low limit alarm, calibration due alarm
- High/low limit alarm, calibration due alarm
- No. of channels:
- 2
- 2
- Calibration:
- pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option
- pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option
- Calibration points:
- 1 point relative mV (RmV) mode, 1 point ORP mode
- 1 point relative mV (RmV) mode, 1 point ORP mode
- W×D×H:
- 240×180×110 mm
- 240×180×110 mm
- Weight:
- 0,9 kg
- 0,9 kg