Miniature circuit breaker lockouts
Supplier: Brady
Certificates
About this item
These miniature circuit breakers are a safe and effective method for locking out European circuit breakers as they fit most of the existing types. Recommended for use in combination with a padlock for added safety.
- Miniature breaker lockout unique in its kind: Requires no tools to install
- Lockout easily installs with the aid of a push button
- Tie-bar style lockout has a convenient thumb wheel for quick application
- Suitable for single and multi-pole breakers
- Made from glass-filled nylon
- One padlock can be applied
Max. shackle Ø: 7 mm
Frequently Bought Together