2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, N-(2,3-Epoxypropyl)phthalimide
CAS Number:5455-98-1
:
Formula:C₁₁H₉NO₃
MDL Number:MFCD00005896
Molecular Weight:203.2 g/mol
Melting Pt:94…100 °C
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:92 to 102 °C
- GC:≥94.0 %
Specification Test Results
Frequently Bought Together