VWR®, Gardner colour standards
Supplier: VWR Chemicals
Certificates
About this item
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis that are designed for grading the colour of similarly coloured liquids, such as resins, varnishes, lacquers, drying oils, fatty acids, lecithin's, sunflower and linseed oil.
- Use either as calibration and/or quality control standards
- Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
- Traceable product certified by an independent laboratory
- Supplied in high quality, tamper-evident bottles
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together