Pierce™ Streptavidin Magnetic Beads
Supplier: Thermo Fisher Scientific
Certificates
About this item
These Pierce™ streptavidin magnetic beads accelerate throughput for automated magnetic purification of biotinylated molecules.
- Streptavidin is immobilised using leach-resistant chemistry
- Superior quality beads with high binding capacity provide rapid and efficient biomolecule purification from complex samples
- Stable, pre-blocked beads provide clean purification products (eg. antigen eluted in IP with biotinylated antibody) that are compatible with mass spectrometry analysis and other downstream analyses
Frequently Bought Together