Order Entry
Czech Republic
ContactUsLinkComponent
Luminometer, BioFix® Lumi-10
Luminometer, BioFix® Lumi-10
  MANA940008
undefined
Luminometer, BioFix® Lumi-10
  MANA940008

 

  • Description:
    Luminometer, BioFix® Lumi-10, 230 V / 50 Hz
  • Environmentally Preferable:
  • Pk:
    1

 

 

BioFix® Lumi-10 is a portable luminometer for the measurement of biological and chemical luminescence reactions with relatively constant light emission.

  • Mobile test system with a high sensitive detector
  • High-resolution graphic display
  • Data memory for up to 2000 test results
  • Variable measurement times to obtain optimum results
  • Serial RS232 interface for data transfer to PC

This device is ideal for environmental analysis/ecological toxicology: acute luminous bacteria tests, hygiene monitoring: ATP and biomass analysis and molecular biological and biochemical diagnostics: reporter gene assays, NADP(H)-measurements, DNA probe assays, luminescence immunoassays.

W×D×H: 150×280×170 mm

Weight: 2 kg (incl. batteries)