ContactUsLinkComponent
Size exclusion chromatography columns, Superdex™ 75 Increase
 :  Cytiva
undefined
Size exclusion chromatography columns, Superdex™ 75 Increase
 :  Cytiva
  • Catalog No:
  • 29-1487-23
  • 29-1487-22
  • 29-1487-21
  • Max. pressure:
  • 20 bar
  • 30 bar
  • 30 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
Frequently Bought Together