(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 97%
Catalog # H63806.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-Chloro-4-methoxybenzylamine hydrochloride
CAS Number:41965-95-1
:
Formula:C₈H₁₁Cl₂NO
:208.09 g/mol
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD01211460
Melting Pt:250…255 °C
ADR:6.1 III