VWR® Prague, Safety Spectacles
Catalog # 111-1846
Supplier: VWR Collection
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Colour:Blue
- Environmentally Preferable:
- Lenses:Clear PC
- Pk:1
Specifications
About this item
These economical, compact cover glasses (OTG) fit over most prescription eyewear.
- Pantoscopic adjustment allows lens angle modification Adjustable temple length
Certifications: 89/686/EEC, EN 166:2001, EN 170; EN 166 FT CE (frame); 1 FT (lens)