- Antibody type:Primary
- Antigen name:Cystinosin, Lysosomal Cystine Transporter
- Antigen symbol:CTNS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:liquid
- Gene ID:1497
- Antigen synonyms:CTNS_HUMAN.|nephropathic|CTNS LSB|Cystinosin|cystinosis|PQLC4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Cystinosis is an autosomal recessive disorder resulting from defective lysosomal transport of cystine and present at birth as a failure to thrive, rickets and proximal renal tubular acidosis. The human CTNS gene on chromosome 17p13 encodes the protein Cystinosin, and mutations in CTNS are responsible for nephropathic cystinosis. The CTNS promoter contains an Sp1 binding element. Cystinosin is an integral membrane protein containing 7 transmembrane domains that functions as a H+-driven transporter responsible for cystine export from lysosomes. In humans, Cystinosin is expressed abundantly in pancreas, kidney (mature and fetal), and skeletal muscle. The mouse homolog to CTNS encodes a protein which is expressed in all tissues except skeletal muscle. In the cell, Cystinosin co-localizes with LAMP-2 to lysosomes. A C-terminal GYDQL sorting motif within Cystinosin is critical for lysosomal localization.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CTNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: