GENbag anaer, generators
Catalog # BMER45534
Supplier: BIOMERIEUX
Specifications
- Description:GENbag anaerobic, 20 sachets
- Environmentally Preferable:
- Pk:20
Specifications
About this item
GENbag consists of an airtight bag made of transparent plastic. Used with a generator sachet, it rapidly produces the appropriate atmosphere for the culture of bacteria which require either an anaerobic, microaerophilic or capnophilic atmosphere. It is used for incubation of single specimens.
- Less than one hour atmosphere requirement
- Adapted to all laboratory requirements
- No hydrogen release thereby making them completely user-safe
- No additional material required