Anti-CASC5 Rabbit Polyclonal Antibody
ABNOPAB21971
: Abnova
- Antibody type:Primary
- Antigen name:Cancer Susceptibility Candidate 5
- Antigen symbol:CASC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:57082
- Antigen synonyms:D40|PPP1R55|Spc7|CT29|hKNL-1|KNL1|AF15Q14|hSpc105
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:HHTEEDIDKSANSVLIKNLSRTPSSCSSSLDSIKADGTSLDFSTYRSSQMESQFLRDTICEESLREKLQDGRITIREFFILLQVHILIQKPRQSNLPGNFTVNTPPTPEDLMLSQYVYRPKIQIYREDCEARRQKIEELKLSASNQDKL
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human CASC5.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant CASC5.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml
Type: Primary
Antigen: CASC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human