Anti-GRM5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16484
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, metabotropic 5
- Antigen symbol:GRM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Hamster,Pig,Rabbit,Horse,Monkey,Chicken,Human,Bovine,Dog,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2915
- Antigen synonyms:GPRC1E|MGLUR5|mGlu5|PPP1R86
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human GRM5.
- Purification:Immunoaffinity purification
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of GRM5.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5.6 ?g/ml
Type: Primary
Antigen: GRM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat