Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cytokeratin 1,2,3,4,5,6,7,8 Mouse Monoclonal Antibody [clone: 10B2315]
Anti-Cytokeratin 1,2,3,4,5,6,7,8 Mouse Monoclonal Antibody [clone: 10B2315]
Catalog # USBIC9097-02
Supplier:  US Biological
Anti-Cytokeratin 1,2,3,4,5,6,7,8 Mouse Monoclonal Antibody [clone: 10B2315]
Catalog # USBIC9097-02
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 1,2,3,4,5,6,7,8
  • Clonality:
    Monoclonal
  • Clone:
    10B2315
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Cytokeratin 1,2,3,4,5,6,7,8
Clonality: Monoclonal
Clone: 10B2315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human