Anti-RNF157 Mouse Monoclonal Antibody [clone: 6B3]
USBI132665
- Antibody type:Primary
- Antigen name:Ring Finger Protein 157
- Antigen symbol:RNF157
- Clonality:Monoclonal
- Clone:6B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:556 to 656
- Storage buffer:PBS, pH 7,2.
- Sequence:PASRAPSEEGEGLPAESPDSNFAGLPAGEQDAEGNDVIEEEDGSPTQEGQRTCAFLGMECDNNNDFDIASVKALDNKLCSEVCLPGAWQADDNAVSRNAQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 0.1ng/ml
Type: Primary
Antigen: RNF157 (ring finger protein 157)
Clonality: monoclonal
Clone: 6B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human