Anti-Ferroportin 1 Rabbit Polyclonal Antibody (Biotin)
NOVUNBP1-21502B
- Antibody type:Primary
- Antigen name:Ferroportin 1
- Antigen symbol:FPN1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:30061
- Antigen synonyms:HFE4|SLC11A3|IREG1|SLC40A1|MST079|solute carrier family 40 member 1|MSTP079|MTP1|FPN1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: Ferroportin 1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse