Anti-KCNH2 Rabbit Polyclonal Antibody
BIRBORB87841-50
: Biorbyt
- Antibody type:Primary
- Antigen name:potassium voltage-gated channel, subfamily H (eag-related), member 2
- Antigen symbol:KCNH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: KCNH2 (potassium voltage-gated channel, subfamily H (eag-related), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human