Anti-GABPB2 Mouse Monoclonal Antibody [clone: 3A3-1F4]
ABNOH00126626-M01
: Abnova
- Antibody type:Primary
- Antigen name:GA Binding Protein Transcription Factor, Beta Subunit 2
- Antigen symbol:GABPB2
- Clonality:Monoclonal
- Clone:3A3-1F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:126626
- Antigen synonyms:BABPB2|beta subuni|E4TF1-53|E4TF1|E4TF1B|NRF2B2|E4TF1-47|beta subunit 1|GA binding protein transcription factor|53kDa|GABPB1|GABPB|beta subunit 2|GABPB2|NRF2B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GABPB2
Clonality: Monoclonal
Clone: 3A3-1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human