Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HMX3 Rabbit Polyclonal Antibody
Anti-HMX3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7424B
Supplier:  Abgent
undefined
Anti-HMX3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7424B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H6 Homeobox 3
  • Antigen symbol:
    HMX3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    MGC32455|Nkx5-2|Homeobox protein Nkx-5.2|Nkx-5.2|Homeobox protein HMX2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: HMX3 (H6 family homeobox 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human