Anti-GNB2L1 Rabbit Polyclonal Antibody
Catalog # AVIVARP40625_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Guanine nucleotide binding protein (G protein), beta 2 like 1
- Antigen symbol:GNB2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10399
- Antigen synonyms:H12.3|Gnb2-rs1|PIG21|HLC-7|RACK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GNB2L1 (guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: