ContactUsLinkComponent
UHPLC columns, Hypersil GOLD™ aQ
UHPLC columns, Hypersil GOLD™ aQ
Certificates
undefined
UHPLC columns, Hypersil GOLD™ aQ
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 554-4383
  • 554-4387
  • 554-4385
  • 554-4649
  • 554-4648
  • 554-4307
  • 554-4386
  • 554-4384
  • Description:
  • 10041-334
  • 10041-348
  • 10041-344
  • 10041-340
  • 10041-328
  • 10041-330
  • 10041-346
  • 10041-342
  • Carbon load:
  • 12%
  • 12%
  • 12%
  • 12%
  • 12%
  • 12%
  • 12%
  • 12%
  • pH range:
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • 2 to 9
  • Surface area:
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g